Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2-M3 Hamster anti-Mouse, Unlabeled, Clone: 130, BD

Catalog No. BDB551769 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551769 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551769 Supplier BD Biosciences Supplier No. 551769

Hamster Monoclonal Antibody

The hamster anti-mouse H2-M3 antibody (clone 130) reacts with the H2-M3 major histocompatibility complex (MHC) non-classical class Ib antigen. H2-M3 (M3) associates with β2-microglobulin and is capable of being expressed by most leukocytes. However, due to a lack of endogenous antigens, M3 has been reported to be undetectable on most cells. Its expression is induced by high-affinity N-formylated peptides from mitochondria, Listeria monocytogenes, and Mycobacterium tuberculosis. The induced expression of M3 is most efficient on antigen
presenting cells. M3 presents antigen to cytotoxic T lymphocytes and may play a key role in protective immunity against the intracellular bacteria L. monocytogenes and M. tuberculosis. Furthermore, M3 is capable of presenting mitochondrial antigens for intrathymic positive selection of T-cell receptors which recognize those peptides.

Host Species: Hamster
Clone: 130
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Purified soluble complex of recombinant truncated M3 and β2-microglobulin

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen H2-M3
Applications Flow Cytometry
Classification Monoclonal
Clone 130
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Purified soluble complex of recombinant truncated M3 and β2-microglobulin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.