Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2A histone family, member V, Mouse, Clone: 2F9, Abnova™

Catalog No. 89015852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-852 Supplier Abnova Corporation Supplier No. H00094239M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant H2AFV.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene encodes a member of the histone H2A family. Several transcript variants encoding different isoforms, have been identified for this gene. [provided by RefSeq

Sequence: MAGGKAGRDSGKAKAKAVSRSQRAGLQFPVGRIHRHLKTRTTSHGRVGATAAVYSAAILEYLTAEVLELAGNASKDLKVKRITPRHLQLAIRGDEELDSLIKATIAGGGVIPHIHKSLIGKKGQQKTA

Specifications

Antigen H2A histone family, member V
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone 2F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant H2AFV.
Formulation PBS with no preservative; pH 7.4
Gene H2AFV
Gene Accession No. BC000098
Gene Alias FLJ26479/H2AV/MGC10170/MGC10831/MGC1947
Gene Symbols H2AFV
Host Species Mouse
Immunogen H2AFV (AAH00098, 1 a.a. ∼ 128 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94239
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.