Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-H2AX; Clone: N1-431; BD
Supplier: BD Biosciences 562377
Description
Host Species: Mouse
Clone: N1-431
Isotype: IgG1, κ
Species Reactivity: Hu, Ms
Immunogen: Phosphorylated Human H2AX Peptide
Flow Cytometry (Intracellular Staining)

Specifications
H2AX | |
N1-431 | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse |
Monoclonal | |
PE | |
Phosphorylated Human H2AX Peptide | |
50 Tests | |
Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction