Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H3 (pS28) Rat, V450, Clone: HTA28, BD

Catalog No. BDB560606 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560606 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560606 Supplier BD Biosciences Supplier No. 560606

Rat Monoclonal Antibody

Histones are highly basic proteins that complex with DNA to form chromatin. Histone H3 is a ∽15kDa protein that is phosphorylated at serine 28 (S28), S10, and/or threonine 11 during mammalian cell mitosis and meiosis. The phosphorylation sites are located in the N-terminal tail, a region that is outside of the chromatin fiber and is thus accessible for interactions with agents that may regulate chromatin or specific gene activities. The phosphorylation states of the two serine sites during the cell cycle are highly regulated by Aurora B kinase and a PP1 phosphatase: S10 is in the phosphorylated state from late G2 phase to anaphase, while S28 is phosphorylated from prophase to anaphase. Furthermore, phosphorylation of histone H3 S28 may be mediated by other kinases in response to external stimuli. Evidence suggests that histone phosphorylation is involved in the regulation of chromosome condensation, cell division, and gene transcription. The HTA28 monoclonal antibody reacts with histone H3 phosphorylated at S28 in its N-terminal tail. It does not recognize the unphosphorylated protein. HTA28 has been predicted to be reactive on human, rat, hamster, and cow samples. Investigators are advised that the BD Hoirzon™ V450 Rat Anti-Histone H3 (pS28) antibody (clone HTA28) is not routinely tested on human, rat, hamster, and cow samples. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen H3 (pS28)
Applications Flow Cytometry
Classification Monoclonal
Clone HTA28
Conjugate Violet Fluor 450 (V450)
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Phosphorylated Human Histone H3 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Hamster, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.