Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HA, Clone: 2E5., Abnova™

Catalog No. 89300355 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-355 Supplier Abnova Corporation Supplier No. MAB5637M40
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen HA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HA (Influenza A virus H1N1).
Formulation In 1x PBS, pH 7.4
Gene Accession No. GQ402204.1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-566 of virus HA (Influenza A virus, A/Canada-MB/RV2013/2009 (H1N1))
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.