Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Epitope Tag (Influenza Hemaglutinin) Chicken anti-Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. AB3254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3254MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3254MI Supplier MilliporeSigma Supplier No. AB3254
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This Anti-HA Epitope Tag (Influenza Hemaglutinin) Antibody, synthetic peptide YPYDVPDYA is validated for use in immunoprecipitation, western blot, IC for the detection of HA Epitope Tag (Influenza Hemaglutinin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Epitope Tag (Influenza Hemaglutinin)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified chicken IgY. Liquid in PBS with 0.02% sodium azide.
Host Species Chicken
Immunogen Synthetic peptide YPYDVPDYA from influenza hemaglutinin (HA) epitope.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.