Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Rabbit anti-All, Clone: 114-2C-7, MilliporeSigma™

Catalog No. 05902RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-902-RMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-902-RMI Supplier MilliporeSigma Supplier No. 05902R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-HA Tag Antibody, clone 114-2C-7 detects level of HA Tag and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Tag
Applications Western Blot
Classification Monoclonal
Clone 114-2C-7
Conjugate Unconjugated
Formulation Protein A purified rabbit immunoglobulin in buffer containing 60mM Tris, 90mM NaCl, 0.03% NaN3, 60mM glycine, pH 7.5, and 40% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide containing the sequence ...CYPYDVPDY... derived from the influenza virus hemaglutinin (HA) protein.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C for 24 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.