Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Rabbit anti-Vertebrates, Clone: DW2, MilliporeSigma™

Catalog No. 04902MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-902-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-902-MI Supplier MilliporeSigma Supplier No. 04902
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-HA Tag Antibody, clone DW2 is an antibody against HA Tag for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Tag
Applications Western Blot
Classification Monoclonal
Clone DW2
Conjugate Unconjugated
Formulation Cultured supernatant in 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence ...CYPYDVPDY... derived from the influenza virus hemaglutinin (HA) protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.