Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ HAF/SART1, Rabbit, Unlabeled, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABC1431
Description
U4/U6.U5 tri-snRNP-associated protein 1 (UniProt O43290; also known as Allergen Hom s 1, HAF, hSART-1, hSnu66, Hypoxia-associated factor, SART-1, SNU66 homolog, Squamous cell carcinoma antigen recognized by T-cells 1, U4/U6.U5 tri-snRNP-associated 110kDa protein) is encoded by the SART1 (also known as HAF) gene (Gene ID 9092) in human. The hypoxia-associated factor (HAF) is an E3 ubiquitin ligase that mediates the ubiquitination and degradation of hypoxia-inducible factor 1alpha (HIF-1alpha). HAF also binds to HIF-2alpha and promotes HIF-2 transactivation instead of promoting HIF-2alpha degradation. In addition, hypoxia-dependent HAF SUMOylation enhances HAF interaction with HIF-2alpha and induction of HIF-2 target genes transcription, such as OCT-3/4, SOX2 and NANOG. HAF therefore promotes stem cell characteristics, aggressive tumor growth and invasion by switching cells from HIF-1alpha- to HIF-2alpha-dependent signaling. HAF overexpression enhances glioblastoma cells that express both HIF-1 and HIF-2alpha to initiate tumors as intracranial xenografts in mice. On the other hand, HAF overexpression results in decreased xenograft tumor growth of HT29 colon carcinoma cells that only express HIF-1alpha.
Specifications
HAF/SART1 | |
Polyclonal | |
Unconjugated | |
SART1 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Target specificity was confirmed by Western blotting analysis of HAF overexpression and HAF shRNA-mediated HAF knockdown in various human cancer cells (Koh, M.Y., et al. (2008). Mol. Cell. Biol. 28(23):7081-7095). | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Immunoprecipitation, Western Blot | |
Please refer to lot specific datasheet. | |
Rabbit polyclonal antibody serum with 0.05% Sodium Azide. | |
O43290 | |
GST-tagged recombinant human HAF/SART1 C-terminal fragment. | |
100 μL | |
Apoptosis & Cancer | |
NP_005137 | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction