Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAGH, Mouse, Clone: 4D3, Abnova™

Catalog No. 89022440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-440 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-440 Supplier Abnova Corporation Supplier No. H00003029M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HAGH.

The enzyme encoded by this gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. [provided by RefSeq

Sequence: GRLPPDTRVYCGHEYTINNLKFARHVEPGNAAIREKLAWAKEKYSIGEPTVPSTLAEEFTYNPFMRVREKTVQQHAGETDPVTTMRAVRREKDQFKMPRD

Specifications

Antigen HAGH
Applications ELISA
Classification Monoclonal
Clone 4D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HAGH.
Formulation PBS with no preservative; pH 7.4
Gene HAGH
Gene Accession No. NM_005326
Gene Alias GLO2/GLX2/GLXII/HAGH1
Gene Symbols HAGH
Host Species Mouse
Immunogen HAGH (NP_005317, 161 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3029
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.