Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

heart and neural crest derivatives expressed 2, Mouse, Clone: 3E3, Abnova™

Catalog No. 89001137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-137 Supplier Abnova Corporation Supplier No. H00009464M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HAND2.

The protein encoded by this gene belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, this transcription factor plays an important role in limb and branchial arch development. [provided by RefSeq

Sequence: LSKIKTLRLATSYIAYLMDLLAKDDQNGEAEAFKAEIKKTDVKEEKRKKELNEILKSTVSSNDKKTKGRTGWPQHVWALELK

Specifications

Antigen heart and neural crest derivatives expressed 2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HAND2.
Formulation PBS with no preservative; pH 7.4
Gene HAND2
Gene Accession No. NM_021973
Gene Alias DHAND2/FLJ16260/Hed/MGC125303/MGC125304/Thing2/bHLHa26/dHand
Gene Symbols HAND2
Host Species Mouse
Immunogen HAND2 (NP_068808.1, 135 a.a. ∼ 216 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9464
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.