Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hyaluronan and proteoglycan link protein 4, Mouse, Clone: 1C1, Abnova™

Catalog No. 89001418 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-418 Supplier Abnova Corporation Supplier No. H00404037M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HAPLN4.

Sequence: QRGRKKVVHVLEGESGSVVVQTAPGQVVSHRGGTIVLPCRYHYEAAAHGHDGVRLKWTKVVDPLAFTDVFVALGPQHRAFGSYRGRAELQGDGPGDASLVLRNVT

Specifications

Antigen hyaluronan and proteoglycan link protein 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HAPLN4.
Formulation PBS with no preservative; pH 7.4
Gene HAPLN4
Gene Accession No. NM_023002
Gene Alias BRAL2/KIAA1926
Gene Symbols HAPLN4
Host Species Mouse
Immunogen HAPLN4 (NP_075378, 30 a.a. ∼ 134 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 404037
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.