Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harmartin (TSC1) Mouse anti-Human, Rat, MilliporeSigma™

Catalog No. 051113MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-111-3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-111-3MI Supplier MilliporeSigma Supplier No. 051113
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Harmartin (TSC1) Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Harmartin (TSC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Harmartin (TSC1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. Q92574
Gene Symbols KIAA0243 , LAM, MGC86987, OTTHUMP00000022439, TSC, hamartin
Host Species Mouse
Immunogen Recombinant protein from human Harmartin
Purification Method Unpurified ascites fluid
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.