Learn More
histidyl-tRNA synthetase, Mouse, Clone: 1C8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HARS.
Supplier: Abnova Corporation H00003035M01
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis. [provided by RefSeq
Sequence: MAERAALEELVKLQGERVRGLKQQKASAELIEEEVAKLLKLKAQLGPDESKQKFVLKTPKGTRDYSPRQMAVREKVFDVIIRCFKRHGAEVIDTPVSpecifications
histidyl-tRNA synthetase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002109 | |
HARS | |
HARS (NP_002100, 1 a.a. ∼ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1C8 | |
Mouse monoclonal antibody raised against a partial recombinant HARS. | |
HARS | |
FLJ20491/HRS | |
Mouse | |
Affinity Purified | |
RUO | |
3035 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.