Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAT1, Polyclonal, Invitrogen™

Catalog No. PIPA125446
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA125446 50 μL
1 options

Catalog No. PIPA125446

Supplier: Thermo Scientific PA125446

Rabbit Polyclonal Antibody

The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene.

PA1-25446 detects HAT1 from human, mouse samples.PA1-25446 has been successfully used in Western blot procedures, where a band of approximately 46kDa is detected. Positive controls: 293 cell extracts, NIH-3T3 cell extracts, and chicken fibroblasts.The PA1-25446 immunogen is a synthetic peptide EESFQELVEDYRRVIERLAQE with N-terminal added lysine conjugated to KLH, corresponding to amino acids 399-419 of Human HAT1.The corresponding sequence differs by three amino acids in chicken and by one amino acid in mouse (putative sequence).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAT1
Applications Western Blot
Classification Polyclonal
Concentration 0.8mg/mL
Conjugate Unconjugated
Gene HAT1
Gene Accession No. O14929, Q8BY71
Gene Alias KAT1
Gene Symbols HAT1
Host Species Rabbit
Immunogen Synthetic peptide EESFQELVEDYRRVIERLAQE with N-terminal added lysine conjugated to KLH, corresponding to amino acids 399-419 of Human HAT1.The corresponding sequence differs by three amino acids in chicken and by one amino acid in mouse (putative sequence).
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8520
Target Species Human, Mouse
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.