Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatitis A virus cellular receptor 1, Mouse, Clone: 2B9, Abnova™

Catalog No. 89001248 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-248 Supplier Abnova Corporation Supplier No. H00026762M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HAVCR1.

Sequence: KVGGEAGPSVTLPCHYSGAVTSMCWNRGSCSLFTCQNGIVWTNGTHVTYRKDTRYKLLGDLSRRDVSLTIENTAVSDSGVYCCRVEHRGWFNDMKITVSL

Specifications

Antigen hepatitis A virus cellular receptor 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HAVCR1.
Formulation PBS with no preservative; pH 7.4
Gene HAVCR1
Gene Accession No. NM_012206
Gene Alias HAVCR/HAVCR-1/KIM-1/KIM1/TIM-1/TIM1/TIMD1
Gene Symbols HAVCR1
Host Species Mouse
Immunogen HAVCR1 (NP_036338, 23 a.a. ∼ 122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 26762
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.