Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HBB, Clone: 7B12, Abnova™

Catalog No. 89300408 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-408 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-408 Supplier Abnova Corporation Supplier No. H00003043M02A

Mouse Monoclonal Antibody

Specifications

Antigen HBB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HBB.
Formulation In ascites fluid
Gene Accession No. BC007075
Gene Alias CD113t-C
Gene Symbols HBB
Host Species Mouse
Immunogen HBB (AAH07075, 38 a.a. ∼ 147 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3043
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.