Learn More
Description
Specifications
Specifications
| Antigen | HCN1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). |
| Gene Symbols | HCN1; HAC-2; BCNG1; BCNG-1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding amino acids 6-24 of rat HCN-1 (Accession number AAF62173). |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.