Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5884200UL
Description
This Anti-HCN1 Antibody is validated for use in western blotting for the detection of HCN1.
Specifications
HCN1 | |
Polyclonal | |
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Western Blot | |
Unconjugated | |
HCN1; HAC-2; BCNG1; BCNG-1 | |
Synthetic peptide corresponding amino acids 6-24 of rat HCN-1 (Accession number AAF62173). | |
200 μL | |
NM_021072.2 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction