Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN3 Rat anti-Mouse, Rat, Clone: TLL 6C5, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB5598
Description
Anti-HCN3 Antibody, clone TLL 6C5 detects level of HCN3 and has been published and validated for use in immunohistochemistry and western blotting.
Specifications
HCN3 | |
Monoclonal | |
Unconjugated | |
NM_020897.1 | |
Rat | |
100 μL | |
Mouse, Rat | |
Supernatant |
Immunohistochemistry, Western Blot | |
TLL 6C5 | |
Cultured supernatant containing 0.05% sodium azide. | |
HCN3, MGC131493, KIAA1535 | |
Synthetic peptide from rat HCN3. | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction