Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HD, Mouse, Clone: 1A12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HD.
Supplier: Abnova Corporation H00003064M13
Description
Sequence: CDGIMASGRKAVTHAIPALQPIVHDLFVLRGTNKADAGKELETQKEVVVSMLLRLIQYHQVLEMFILVLQQCHKENEDKWKRLSRQIADIILPMLAKQQMHIDSSpecifications
HD | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002111 | |
HTT | |
HD (NP_002102, 1524 a.a. ∼ 1627 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
1A12 | |
Mouse monoclonal antibody raised against a partial recombinant HD. | |
HTT | |
HD/IT15 | |
Mouse | |
Affinity chromatography | |
RUO | |
3064 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction