Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HD, Mouse, Clone: 3D6, Abnova™

Catalog No. 89022461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-461 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-461 Supplier Abnova Corporation Supplier No. H00003064M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HD.

Sequence: AVAEEPLHRPKKELSATKKDRVNHCLTICENIVAQSVRNSPEFQKLLGIAMELFLLCSDDAESDVRMVADECLNKVIKALMDSNLPRLQLELYKEIKKNGAPRSLRAALW

Specifications

Antigen HD
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HD.
Formulation PBS with no preservative; pH 7.4
Gene HTT
Gene Accession No. NM_002111
Gene Alias HD/IT15
Gene Symbols HTT
Host Species Mouse
Immunogen HD (NP_002102, 81 a.a. ∼ 190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3064
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.