Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE260MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE260MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE260MI Supplier MilliporeSigma Supplier No. ABE260

Rabbit Polyclonal Antibody

Anti-HDAC1 Antibody is a Rabbit Polyclonal Antibody for detection of HDAC1 also known as Histone deacetylase 1, HD1 and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HDAC1; RPD3L1
Host Species Rabbit
Immunogen Linear Peptide corresponding to human HDAC1.
Purification Method Affinity Purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_004955
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.