Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC11 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071548MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-154-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-154-8MI Supplier MilliporeSigma Supplier No. 071548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-HDAC11 Antibody (Rabbit Polyclonal Antibody) validated in western blot, ELISA, immunohistochemistry to detect HDAC11 also known as histone deacetylase 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC11
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.09% NaN3.
Gene Symbols HD11 ; FLJ22237 ; EC 3.5.1.98
Host Species Rabbit
Immunogen The immunogen was a KLH-conjugated synthetic peptide corresponding the N-terminal region of human HDAC11.
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_079103
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.