Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC2 Mouse anti-Human, Bovine, Mouse, Rat, Hamster, Clone: 3F3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05814
Description
Anti-HDAC2 Antibody, clone 3F3 is a Mouse Monoclonal Antibody for detection of HDAC2 also known as YY1-associated factor 1, histone deacetylase 2 and has been validated in immunocytochemistry, immunoprecipitation and western blotting.
Specifications
| HDAC2 | |
| Monoclonal | |
| Unconjugated | |
| NP_001518 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C for up to 12 months. | |
| IgG1 |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 3F3 | |
| Purified mouse monoclonal IgG in buffer containing 70% storage buffer (0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide) and 30% glycerol. Store at −20°C. | |
| HDAC2, HD2, YAF1, RPD3 | |
| KLH-conjugated, synthetic peptide (CEKTDTKGTKSEQLSNP) corresponding to amino acids 473-488 of human HDAC2 (Accession # NP_001518) containing a N-terminal cysteine | |
| 200 μg | |
| Bovine, Hamster, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction