Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06890MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-890-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-890-MI Supplier MilliporeSigma Supplier No. 06890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-HDAC3 Antibody is a Rabbit Polyclonal Antibody for detection of HDAC3 also known as histone deacetylase 3 and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols HDAC3; SMAP45; HD3; RPD3; RPD3-2
Host Species Rabbit
Immunogen peptide corresponding to amino acids 411-428 of human HDAC3 (NEFYDGDHDNDKESDVEI)
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003883
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.