Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Mouse anti-Bovine, Human, Hamster, Mouse, Clone: 3G6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05813
Description
Anti-HDAC3 Antibody, clone 3G6 is a Mouse Monoclonal Antibody for detection of HDAC3 also known as histone deacetylase 3 and has been tested in immunoprecipitation, western blot, immunocytochemistry.
Specifications
| HDAC3 | |
| Monoclonal | |
| Unconjugated | |
| NM_003883 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C for 24 months. | |
| IgG2a |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 3G6 | |
| 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%. | |
| HDAC3, SMAP45, HD3, RPD3, RPD3-2 | |
| peptide (NEFYDGDHDNDKESDVEI) corresponding to amino acids 411-428 of human HDAC3 | |
| 200 μg | |
| Bovine, Hamster, Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction