Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC4 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE262MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE262MI Supplier MilliporeSigma Supplier No. ABE262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-HDAC4 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry, immunoprecipitation, Chimmunoprecipitation to detect HDAC4 also known as Histone deacetylase 4, HD4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC4
Applications ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols HDAC4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human HDAC4.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_006028
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.