Learn More
HDAC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA131473
Description
PA1-31473 detects HDAC6 from rat, mouse, human samples. PA1-31473 has been successfully used in Chimmunoprecipitation, immunohistochemistry (paraffin tissue), Western blot applications. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene belongs to class II of the histone deacetylase/acuc/apha family. It contains an internal duplication of two catalytic domains which appear to function independently of each other. This protein possesses histone deacetylase activity and represses transcription.
Specifications
HDAC6 | |
Polyclonal | |
Unconjugated | |
Q9UBN7, Q9Z2V5 | |
HDAC6 | |
Proprietary immunogen | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
HDAC6 | |
FLJ16239, HD6, JM21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10013, 15185, 84581 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.