Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC8 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07505MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-505-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-505-MI Supplier MilliporeSigma Supplier No. 07505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-HDAC8 Antibody is a Rabbit Polyclonal Antibody for detection of HDAC8 also known as histone deacetylase 8 and has been tested in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC8
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 30% glycerol
Gene Symbols HDAC8; HD8; RPD3; HDACL1
Host Species Rabbit
Immunogen GST fusion protein corresponding to amino acids 305-377 of human HDAC8
Purification Method Antiserum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_018486
Target Species Human, Mouse
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.