Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC9 Rat anti-Human, Clone: LH/JC2, MilliporeSigma™

Catalog No. 05897MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-897-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-897-MI Supplier MilliporeSigma Supplier No. 05897

Rat Monoclonal Antibody

Detect HDAC9 also known as histone deacetylase 9 with Anti-HDAC9 Antibody, clone LH/JC2 (Rat Monoclonal Antibody), that has been demonstrated to work in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC9
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone LH/JC2
Conjugate Unconjugated
Gene Accession No. NM_014707.1; NM_058176.2; NM_058177.2; NM_178423.1; NM_178425.2
Gene Symbols HDAC9, HDAC9FL, MITR, HD9, HDRP, HDAC9B, KIAA0744, DKFZp779K1053, HDAC, HD7B, HD7, HDAC7B, HDAC7
Host Species Rat
Immunogen synthetic peptide corresponding to amino acids 1046-1060 of human HDAC9 isoform 5
Purification Method Protein G purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.