Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Factor I Rat anti-Human, Mouse, Monkey, Clone: 10H8, MilliporeSigma™

Catalog No. MAB88078MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB88078MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB88078MI Supplier MilliporeSigma Supplier No. MAB88078

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

This Anti-Heat Shock Factor I Antibody, clone 10H8 is validated for use in ELISA, immunofluoresence, immunoprecipitation, western blotting for the detection of Heat Shock Factor I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heat Shock Factor I
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10H8
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 10mM PBS, pH 7.4 with 0.2% BSA and 15mM sodium azide.
Gene Accession No. NM_005526.2
Gene Symbols HSF1, HSTF1
Host Species Rat
Immunogen Full length recombinanat mouse HSF1 protein (aa 1-503).
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Monkey, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.