Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Protein 70 Mouse anti-Bovine, Human, Monkey, Clone: W27, MilliporeSigma™

Catalog No. MAB3516MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3516MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3516MI Supplier MilliporeSigma Supplier No. MAB3516
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Heat Shock Protein 70 Antibody, clone W27 is validated for use in immunoprecipitation, western blot, immunohistochemistry (paraffin) for the detection of Heat Shock Protein 70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heat Shock Protein 70
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone W27
Conjugate Unconjugated
Formulation Liquid in 10mM PBS, pH 7.4 with 0.2% BSA and 15mM sodium azide.
Gene Accession No. NM_198431.1; NM_002154.3
Gene Symbols HSPA4, APG2, hsp70, HSP70RY, HS24/P52, RY, MGC131852, APG-2, hsp70RY
Host Species Mouse
Immunogen HSP70 from HeLa cells.
Quantity 500 μL
Primary or Secondary Primary
Target Species Bovine, Human, Monkey
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.