Learn More
anti-Heat Shock Protein 75/HSP75/ TRAP-1 Ab-2, Clone: TR-1A, Epredia™ LabVision
Shop All Epredia ProductsDescription
A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (HSP's), more appropriately called as stress response proteins (SRP's). The Hsp90 family of proteins in mammalian cells consists of Hsp90 α and β, Grp94, and Trap-1 (Hsp75). It was found that human TRAP-1 is localized to mitochondria and exhibited ATPase activity that was inhibited by both geldanamycin and radicicol indicating that TRAP-1 has functions that are distinct from those of hsp90. It was also found that TRAP-1 had a biochemical activity to refold denatured retinoblastoma protein (Rb) into its native conformation.
Host Species: Mouse
Clone: TR-1A
Isotype: IgG2a
Cross Reactivity: Human. Others-not known.
Epitope: Not determined
Immunogen: Recombinant TRAP-1 protein
Molecular Weight: 75kDa
Positive Control: HeLa, SKBR-3 or A431 cells
Cellular Localization: cytoplasmic
Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Heat Shock Protein 75/HSP75/ TRAP-1 Ab-2 |
| Classification | Monoclonal |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant TRAP-1 protein |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.