Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Heat Shock Protein 75/HSP75/ TRAP-1 Ab-2, Clone: TR-1A, Epredia™ LabVision

Catalog No. MS1309PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
250μg/100μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1309PABX 200 μL 1mg/mL
MS1309P0 100 μL 200μg/mL
MS1309P1 500 μL 200μg/mL
MS1309P 1 mL 200μg/mL
MS1309PCL 100 μL 250μg/100μL
MS1309P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1309PABX Supplier Epredia Supplier No. MS1309PABX

Mouse Monoclonal Antibody

A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (HSP's), more appropriately called as stress response proteins (SRP's). The Hsp90 family of proteins in mammalian cells consists of Hsp90 α and β, Grp94, and Trap-1 (Hsp75). It was found that human TRAP-1 is localized to mitochondria and exhibited ATPase activity that was inhibited by both geldanamycin and radicicol indicating that TRAP-1 has functions that are distinct from those of hsp90. It was also found that TRAP-1 had a biochemical activity to refold denatured retinoblastoma protein (Rb) into its native conformation.

Host Species: Mouse
Clone: TR-1A
Isotype: IgG2a
Cross Reactivity: Human. Others-not known.
Epitope: Not determined
Immunogen: Recombinant TRAP-1 protein
Molecular Weight: 75kDa
Positive Control: HeLa, SKBR-3 or A431 cells
Cellular Localization: cytoplasmic

Immunoprecipitation, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Heat Shock Protein 75/HSP75/ TRAP-1 Ab-2
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant TRAP-1 protein
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.