Learn More
Heat Shock Protein 90Β Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3468
Description
This Anti-Heat Shock Protein 90β Antibody is validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry (paraffin) for the detection of Heat Shock Protein 90β.
Specifications
Heat Shock Protein 90β | |
Polyclonal | |
Purified immunoglobulin. Liquid in 10mM PBS, pH 7.4 with 0.2% BSA and 15mM sodium azide. | |
Rabbit | |
500 μL | |
NM_007355.2 | |
2-8°C in undiluted aliquots for up to 6 months |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
HSP90AB1; HSP90-BETA; HSPC2; FLJ26984; D6S182; HSP90B; OTTHUMP00000039869; HSPCB | |
Synthetic peptide corresponding to amino acids 2-13 from the N-terminus of mouse heat shock protein 90b. | |
Primary | |
Human, Mouse, Rat | |