Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Protein 90α Rabbit anti-Human, Mouse, Rat, Canine, Polyclonal, MilliporeSigma™

Catalog No. 072174MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-217-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-217-4MI Supplier MilliporeSigma Supplier No. 072174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect the Heat Shock Protein 90α protein using this Anti-Heat Shock Protein 90α validated for use in western blot, immunoprecipitation, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heat Shock Protein 90α
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HSP90AA1; HSPCAL4; HSPN; HSP86; Hsp89; HSP90N; HSPC1; HSP90A; Hsp90; HSPCAL1; HSPCA; LAP2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Heat Shock Protein 90α.
Purification Method Protein A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001017963
Target Species Canine, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.