Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90 alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA129139
Description
PA1-29140 detects p107 from mouse, human, rat samples. PA1-29140 has been successfully used in Western blot applications. The PA1-29140 immunogen is: Synthetic peptide corresponding to C-terminus of human p107 protein
Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two serperate gene products, HSP90 alpha and HSP90 beta. This antibody weakly cross reacts with hsp90Beta/hsp84.
Specifications
HSP90 alpha | |
Polyclonal | |
Unconjugated | |
P07900, P07901, P82995 | |
HSP90AA1 | |
Synthetic 11-mer peptide corresponding to residues 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90 alpha. | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
HSP90AA1 | |
HSP86; HSP89A; HSP90A; HSP90N; HSPC1; HSPCA; HSPCAL1; HSPCAL4; HSPN; Hsp89; Hsp90; LAP2; HSP 86; heat shock 86 kDa; heat shock 90kD protein 1; alpha; heat shock 90kD protein 1; alpha-like 4; heat shock 90kD protein; alpha-like 4; heat shock 90kDa protein 1; alpha; heat shock protein HSP 90-alpha; renal carcinoma antigen NY-REN-38 | |
Rabbit | |
Protein A | |
RUO | |
15519, 299331, 3320 | |
4° C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction