Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

heme binding protein 1 (B02P), Mouse anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017073 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-073 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-073 Supplier Abnova Corporation Supplier No. H00050865B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HEBP1 protein.

The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells. [provided by RefSeq

Sequence: MLGMIKNSLFGSVETWPWQVLSKGDKEEVAYEERACEGGKFATVEVTDKPVDEALREAMPKVAKYAGGTNDKGIGMGMTVPISFAVFPNEDGSLQKKLKVWFRIPNQFQSDPPAPSDKSVKIEEREGITVYSMQFGGYAKEADYVAQATRLRAALEGTATYRGDIYFCTGYDPPMKPYGRRNEIWLLKT

Specifications

Antigen heme binding protein 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HEBP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene HEBP1
Gene Accession No. NM_015987.3
Gene Alias HBP/HEBP
Gene Symbols HEBP1
Host Species Mouse
Immunogen HEBP1 (AAH16277.1, 1 a.a. ∼ 189 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50865
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.