Learn More
HEC1 Goat anti-Human, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA126543
Description
PA1-26543 detects HEC1 from human samples. PA1-26543 has been successfully used in Western blot procedures. The PA1-26543 immunogen is a synthetic peptide corresponding to residues Y E K K A T L I K S S E E of HEC1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
HEC is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Specifications
HEC1 | |
Polyclonal | |
Unconjugated | |
O14777 | |
NDC80 | |
Synthetic peptide corresponding to residues Y E K K A T L I K S S E E of HEC1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
0.5 mg/mL | |
NDC80 | |
HEC, HEC1, KNTC2, TID3, hsNDC80 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.