Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helios Armenian Hamster anti-Human, Alexa Fluor 647, Clone: 22F6, BD

Catalog No. BDB563951 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563951 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563951 Supplier BD Biosciences Supplier No. 563951
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The 22F6 monoclonal antibody specifically binds to mouse and human Helios. Helios is a member of the Ikaros family of zinc-finger transcription factors, which play important roles in hematopoietic cell development and tumor suppression. Helios expression is restricted to the earliest stages of embryonic hematopoiesis, a variety of epithelial tissues and is notably increased in thymic-derived regulatory CD4+Foxp3+ T (Treg) cells. Its high expression levels in Treg cells are independent from Foxp3 and are believed to contribute, along with other transcription factors, to the phenotypic stability of natural regulatory T cells. Accordingly, it has been demonstrated that Helios directly stimulates Foxp3 transcription while it inhibits ll2 gene expression, contributing for the maintenance of cellular anergy. Helios is also differentially expressed during negative and positive selection in the thymus, marking CD4+ autoreactive cells for deletion. Helios may possibly play roles in T cell activation, since it is upregulated in Th2 and Tfh cells. Despite these roles in T cell development and function, Helios genetic ablation in mice revealed no significant abnormalities in Treg or other T cell subsets. This finding suggests that other Ikaros family members may play redundant roles.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Helios
Applications Flow Cytometry
Classification Monoclonal
Clone 22F6
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description IKAROS family zinc finger protein 2; ANF1A2; HELIOS; ZNF1A2; ZNFN1A2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Armenian Hamster
Immunogen Mouse Helios Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.