Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematopoietic progenitor cell Mouse anti-Human, PE, Clone: BB9, BD

Catalog No. BDB557928 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557928 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557928 Supplier BD Biosciences Supplier No. 557928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody BB9 recognizes a cell-surface glycoprotein of approximately 160kDa, expressed on primitive hematopoietic progenitor cells (HPC). This antibody was generated using human bone marrow-derived stromal cells as immunogen. BB9 reacts with stromal cells, a minor subset of human bone marrow cells; it does not react with peripheral blood leukocytes. Reacts with a subset of CD34+, CD90+ demonstrating low or absent expression of CD38, low retention of Rhodamine 123 and Ki-67-. CD34+BB9+ cells are able to sustainm hematopoiesis in pre-CFU culture stimulated with IL-3, IL-6, G-CSF and SCF. BB9 also reacts with a subset of CD34+ cells from mobilized peripheral blood.

Host Species: Mouse
Clone: BB9
Isotype: IgG1, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hematopoietic progenitor cell
Applications Flow Cytometry
Classification Monoclonal
Clone BB9
Conjugate PE
Description ACE1; DCP1; Kininase II
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human bone marrow-derived stromal cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.