Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heme Oxygenase 1 Mouse anti-Human, Unlabeled, Clone: 23, BD

Catalog No. BDB610713 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610713 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610713 Supplier BD Biosciences Supplier No. 610713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Heme oxygenases 1 and 2 (HO-1, HO-2) cleave the heme molecule, resulting in the production of carbon monoxide (CO) and biliverdin. HO-1 is a 288 amino acid monooxygenase with a molecular weight of 32kDa. While HO-2 is constitutively expressed in tissues, HO-1 is rapidly induced by several stimuli such as lutathione depletion, hemin, heat shock, heavy metals, oxidative stress, oxidized LDL, anoxia, and endotoxic shock. Similar to nitric oxide (NO), CO activates guanylate cyclase and reduces platelet aggregation. Several conditions that increase NO also induce HO-1 expression and activity. This suggests that NO synthases, Cox-2, and HO-1 are part of a general defense mechanism against stress and pathogen invasion.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Heme Oxygenase 1
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Heme Oxygenase 1 (HO-1) aa. 150-286
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.