Learn More
Heme Oxygenase 1 Rabbit anti-Bovine, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Heme Oxygenase 1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Liquid in PBS containing 0.01M sodium azide. |
| Gene Symbols | HMOX1; HO; OTTHUMP00000028925; HO1; HO-1; bK286B10 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide with the amino acid sequence H-D-L-S-E-A-L-K-E-A-T-K-E-V-H-C-NH2 corresponding to amino acid residues [12-25] + Cys-NH2 of the human HO-1 protein (Yoshida et al., 1988). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.