Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HEMK2, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00029104B01P
Specifications
HEMK2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HEMK2 protein. | |
N6AMT1 | |
C21orf127/HEMK2/MGC19995/MTQ2/N6AMT/PRED28 | |
Mouse | |
Affinity chromatography | |
RUO | |
29104 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011554.1 | |
N6AMT1 | |
HEMK2 (AAH11554.1, 1 a.a. ∼ 186 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction