Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heparan Sulfate Proteoglycan (Perlecan) Mouse anti-Human, Clone: 5D7-2E4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT12
Description
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone 5D7-2E4 detects level of Heparan Sulfate Proteoglycan (Perlecan) and has been published and validated for use in immunohistochemistry.
Specifications
Heparan Sulfate Proteoglycan (Perlecan) | |
Monoclonal | |
Unconjugated | |
NP_005520 | |
Mouse | |
Protein G | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry | |
5D7-2E4 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HSPG2, SJA, perlecan, SJS, HSPG, PRCAN, Perlecan, SJS1, PLC | |
Full length recombinant protein corresponding to human Perlecan. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction