Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparan Sulfate Proteoglycan (Perlecan) Rat anti-Human, Clone: A7L6, MilliporeSigma™

Catalog No. MAB1948PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1948PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1948PMI Supplier MilliporeSigma Supplier No. MAB1948P
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Heparan Sulfate Proteoglycan (Perlecan) using this Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone A7L6 validated for use in immunohistochemistry, immunocytochemistry, western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heparan Sulfate Proteoglycan (Perlecan)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A7L6
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005520
Gene Symbols HSPG2, SJS, PRCAN, SJS1, SJA, perlecan, HSPG, Perlecan, PLC
Host Species Rat
Immunogen Heparan Sulfate Proteoglycan from EHS mouse tumor
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.