Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis A Virus Mouse anti-Human, MilliporeSigma™

Catalog No. MAB8241MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8241MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8241MI Supplier MilliporeSigma Supplier No. MAB8241
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Hepatitis A Virus Antibody detects level of Hepatitis A Virus and has been published and validated for use in ELISA, immunofluoresence, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis A Virus
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.01Mphosphate buffered saline (PBS), pH 7.2 containing 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.