Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Hepatitis A Virus Virions Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA173089
Description
The PA1-73089 antibody reacts with Hepatitis A virions. PA1-73089 has been successfully used in ELISA applications. The PA1-73089 antibody was raised against Hepatitis A virus strain HM175.
Hepatitis A virus (HAV), a human picornavirus, is spread by the fecal-oral route, but the site of virus replication is the liver. The symptoms of the disease are heterogeneous depending on the age of infection, ranging from silent infections mainly in young children to classical hepatitis in older age groups, and fatal courses of the disease also occur.
Specifications
Hepatitis A Virus Virions | |
Polyclonal | |
Unconjugated | |
HAVgp1; HepA; hepatitis a virus surface antigen; hepatitis a virus vp1; hepatitis a virus vp4 | |
Purified virions of strain HM175. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG Fraction | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction