Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Mouse anti-Human, Clone: 146, MilliporeSigma™

Catalog No. MAB8429MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8429MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8429MI Supplier MilliporeSigma Supplier No. MAB8429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Hepatitis B Virus using this Anti-Hepatitis B Virus Antibody, X-Protein, a.a. 50-88, clone 146 validated for use in ELISA, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 146
Conjugate Unconjugated
Formulation Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Host Species Mouse
Immunogen Peptide cooresponding to amino acids 50-88 of human HBV-X protein {gahlslrglpvcafssagpca lrltsvrrmettvnahqv}
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.